Learn More
Kemtech America™ Synthware™ Filter Tubes with Fritted Disc
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Manufacturer:  Kemtech America™ F702440C
 View more versions of this product
Catalog No. 31-501-560
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Filter tube with fritted disc
Approximate Frit O.D.: 36mm, Coarse porosity
24/40