Kemtech America™ Synthware™ Gas Dispersion Tubes, Fritted, 15° Angle

For use with filtration techniques

Manufacturer: Kemtech America™ F273035X

 View more versions of this product

Catalog No. 31-501-549

  Check Availability
Add to Cart

    Specifications

    Description Gas dispersion tube, fritted, 15° Angle
    Diameter (Metric) Inner Disc: 30mm
    Item Description Porosity: Extra Coarse

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit