Kemtech America™ Synthware™ Liebig Condenser
With standard taper outer joint at the top and lower inner drip tip joint
Manufacturer: Kemtech America™ C209180
 View more versions of this product
Catalog No. 31-500-811
  Check Availability
Add to Cart
Related Products
Product Content Feedback