Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Liebig Condenser, with Removable Hose Connections
GREENER_CHOICESDP

Condenser has a standard taper outer joint at the top and a lower inner drip tip joint

Supplier:  Synthware™ C509300

 View more versions of this product

Catalog No. 31-500-804


Add to Cart

Description

Description

  • Joints of same size
Specifications

Specifications

19/22
Liebig Condenser
Hose connection: 8mm O.D.
Glass
300 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit