Learn More
Synthware™ Reaction Tubes with High Vacuum Valve
Cylindrical shaped tube used to connect two or more glass pieces
Supplier: Synthware™ F564250
Description
- With standard taper outer joint and side arm with high vacuum valve
- Valves with PTFE O-ring at tip and PTFE wiper to protect Viton O-ring on shaft
Specifications
Reaction Tube with High Vacuum Valve | |
250 mL, 0-4 mm | |
24/40 |