Learn More
Synthware™ Reflux Condenser, with Removable Hose Connections
For use with volatile solvents
Supplier: Synthware™ C561100
Description
- Coil style reflux condenser with tightly wrapped coil providing maximum cooling
- With top outer standard taper joint and lower inner drip tip joint
Specifications
Glass | |
100 mm | |
Reflux Condenser |
210 mm | |
Hose connection: 8mm O.D. |