Learn More
Synthware™ Round Bottom Flasks with Hooks
Short neck with standard taper outer joint
Supplier: Synthware™ F331450
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
- Includes two hooks that are 180° apart
Specifications
Short neck round bottom flask | |
50 mL | |
14/20 |