Learn More
Synthware™ Separatory Funnels with PTFE Stopcock and Stopper
Includes PTFE Stopper
Supplier: Synthware™ F451050
Description
- Upper outer and lower inner standard taper joints are same size
Specifications
50 mL | |
14/20 | |
PTFE |
Glass | |
2mm | |
Separatory funnel |