Kemtech America™ Synthware™ Solvent Addition Funnels

Permits addition of solvents to flash chromatography columns without disturbing silica-gel/packing layer

Manufacturer: Kemtech America™ F292810

 View more versions of this product

Catalog No. 31-501-485

  Check Availability
Add to Cart

    Specifications

    Joint Number Fits 45/50 or 50/30
    Product Type Solvent addition funnel
    Diameter (Metric) Outer Stem 28mm
    Diameter (Metric) Top 90mm
    Length (Metric) Stem 100mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit