Learn More
Synthware™ Solvent Transfer/Storage Flasks
For long term deoxygenated solvent storage
Supplier: Synthware™ F484258
Description
- Center neck joint sealed off from direct access to flask
- A syringe or cannula is used to recover the solvent through valve
Specifications
Valve Size: 0-8mm; Solvent storage/transfer flask | |
250 mL | |
24/40 |