Learn More
Synthware™ Storage Flasks with Side Arm
Supplied with high vacuum valve with PTFE O-ring at tip
Supplier: Synthware™ F461025
Description
SYNTHWARE storage flask with standard taper outer joint and high vacuum valve. Valve has a PTFE O-ring at the tip. Valve is used to control liquid or gas flow into the flask.Specifications
Clear | |
25 mL | |
Organic Synthesis |
Storage flask with outer joint, Valve Size: 0-4mm | |
Borosilicate Glass (3.3) | |
14/20 |