Kemtech America™ Synthware™ Storage Tubes with High Vacuum Valve

For long term deoxygenated solvent storage

Manufacturer: Kemtech America™ F580100

 View more versions of this product

Catalog No. 31-501-339

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 100mL
    Diameter (Metric) Outside Tube: 40mm
    Item Description Valve Size: 0-4mm
    Length (Metric) Tube: 165mm
    Product Type Stotage tube with high vacuum valve

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit