Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtech America™ Synthware™ Storage Tubes with High Vacuum Valve

For long term deoxygenated solvent storage

Manufacturer:  Kemtech America™ F580025

 View more versions of this product

Catalog No. 31-501-335


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

25mL
Valve Size: 0-4mm
Stotage tube with high vacuum valve
Tube: 26mm
Tube: 110mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit