Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Three Neck Flask, Angled
Three Neck Flask with standard taper outer joints on both center and angled side necks.
Supplier: Synthware™ F431500
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness.
Specifications
Three Neck Round Bottom Flasks, Angled | |
Borosilicate Glass (3.3) | |
Center: 14/20, Side: 14/20 | |
Reusable | |
Beaded | |
Round |
500 mL | |
Three-neck Flask | |
Autoclavable | |
No | |
Non-sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction