Learn More
Synthware™ Three Neck Flask, Angled
GREENER_CHOICESDP
Three Neck Flask with standard taper outer joints on both center and angled side necks.
Supplier:  Synthware™ F431045
 View more versions of this product
Catalog No. 31-501-262
Add to Cart
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness.
Specifications
Specifications
Three Neck Round Bottom Flasks, Angled
Borosilicate Glass (3.3)
Center: 45/50, Side: 24/40
Reusable
Beaded
Round
1 L
Three-neck Flask
Autoclavable
No
Non-sterile
Product Suggestions
Product Suggestions