Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtech America™ Synthware? Vacuum/Inert Gas Adapters, 90°, with Outer Joint

Allows flexible tubing connection for introduction of vacuum or inert gas to reaction system

Manufacturer:  Kemtech America™ A951420

 View more versions of this product

Catalog No. 31-500-543


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Vacuum/Inert Gas Adapter, 90°, with Outer Joint
14/20
Hose O.D.: 8mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit