Kemtech America™ Synthware™ West Condenser, with Removable Hose Connections
Has narrow annular space between the inner tube and jacket that provides high cooling efficiency
Manufacturer: Kemtech America™ C551110
 View more versions of this product
Catalog No. 31-500-769
  Check Availability
Add to Cart
Related Products
Product Content Feedback