Learn More
Thermo Scientific Chemicals Syntide 2, 96%
For research use
$95.65 - $371.65
Specifications
For Use With (Application) | Enzyme Assay Substrate |
---|---|
Formulation | Lyophilized powder |
Storage Requirements | -30° to -10°C |
Regulatory Status | RUO |
Protein Length | Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ62843MCR
|
Thermo Scientific Chemicals
J62843MCR |
1mg |
Each
for $95.65
Save 8% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ62843LB0
|
Thermo Scientific Chemicals
J62843LB0 |
5mg |
Each
for $371.65
Save 8% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Selective substrate for calmodulin-dependent protein kinase II
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Enzyme Assay Substrate | |
-30° to -10°C | |
Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys | |
Native | |
96% |
Lyophilized powder | |
RUO | |
Syntide 2, 96% | |
Lyophilized |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.