Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntrophin alpha 1 Mouse anti-Human, Alexa Fluor™ 750, Clone: 10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222587AF750

Catalog No. NP22287A70


Add to cart

Description

Description

Syntrophin alpha 1 Monoclonal antibody specifically detects Syntrophin alpha 1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Syntrophin alpha 1
10
Western Blot, ELISA
Conjugated, Purified
SNTA1
Recombinant human SNTA1 (1-505aa) purified from E. coli
Protein A purified
Cytoskeleton Markers, Neuroscience, Vision
Primary
6640.0
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit