Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntrophin beta 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Syntrophin beta 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15978420
|
Novus Biologicals
NBP15978420UL |
20 μL |
Each for $152.22
|
|
NBP159784
|
Novus Biologicals
NBP159784 |
100 μL |
Each for $436.00
|
|
Description
Syntrophin beta 1 Polyclonal specifically detects Syntrophin beta 1 in Human samples. It is validated for Western Blot.Specifications
Syntrophin beta 1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
beta-1-syntrophin, 59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, syntrophin, beta 1 (dystrophin-associated protein A1, 59kDa, basic component 1), TIP-43 | |
SNTB1 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
Q13884 | |
6641 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title