Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Synuclein Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1572P

 View more versions of this product

Catalog No. MS1572P


Add to cart

Description

Description

Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.

Host Species: Mouse

Clone: syn211

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant α-synuclein

Molecular Weight: ~14kDa

Positive Control: Brain

Cellular Localization: Neuronal cytosol and presynaptic terminals

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Synuclein Ab-2
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
syn211
Unconjugated
Concentrated
Human recombinant alpha-synuclein
1mL
Neuroscience
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit