Learn More
Synuclein Ab-2 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results Epredia™ Synuclein Ab-2, Mouse Monoclonal Antibody.
$735.00 - $1196.00
Specifications
Antigen | Synuclein Ab-2 |
---|---|
Clone | syn211 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1572PABX
|
Epredia
MS1572PABX |
Concentrated | 200 μL |
Each for $1,196.00
|
|
||||
MS1572P1
|
Epredia
MS1572P1 |
Concentrated | 500 μL |
Each for $735.00
|
|
||||
MS1572P0
|
Epredia
MS1572P0 |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1572P1ABX
|
Epredia
MS1572P1ABX |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1572R1
|
Epredia
MS1572R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.Host Species: Mouse
Clone: syn211
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: Human recombinant α-synuclein
Molecular Weight: ∽14kDa
Positive Control: Brain
Cellular Localization: Neuronal cytosol and presynaptic terminals
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Synuclein Ab-2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG1 |
syn211 | |
Monoclonal | |
Mouse | |
Neuroscience | |
Human recombinant alpha-synuclein | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.