Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ α-Synuclein Rabbit Polyclonal Antibody

Synuclein α, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$1104.00

Specifications

Antigen Synuclein alpha
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
RB9026P
View Documents
Thermo Scientific™
RB9026P
Concentrated 1mL
Each for $1,104.00
Add to cart
 
RB9026R1
View Documents
Thermo Scientific™
RB9026R1
Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
RB9026P0
View Documents
Thermo Scientific™
RB9026P0
Concentrated 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9026P1
View Documents
Thermo Scientific™
RB9026P1
Concentrated 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9026R7
View Documents
Thermo Scientific™
RB9026R7
Prediluted 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Synuclein α, Rabbit Polyclonal Antibody.

α-synuclein is a protein of 140 amino acids and a member of the synuclein family. It may be implicated in the formation of Lewy bodies and the selective degeneration of neurons in sporadic Parkinson's disease and Lewy body dementia. This antibody is specific for α-synuclein and does not cross react with β-synuclein and will be useful for the confirmation of Parkinson's disease and Lewy body dementia and other studies for the localization and detection of α-synuclein.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide derived from near C-terminus of human α-Synuclein

Molecular Weight: 15kDa

Positive Control: Posterior cingulated gyrus

Cellular Localization: Neuronal cytosol and presynaptic terminals

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Synuclein alpha
Polyclonal
Rabbit
Neuroscience
A synthetic peptide derived from near C-terminal of human alpha Synuclein
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit