Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYPL1 Mouse anti-Human, Clone: EP10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259340

Catalog No. NBP259340


Add to cart

Description

Description

SYPL1 Monoclonal antibody specifically detects SYPL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

SYPL1
EP10
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Purified
SYPL1
Immunoprecipitate of human brain
Protein G purified
RUO
Primary
6856
Human
Western Blot, Immunohistochemistry
Unconjugated
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Detects 38kDa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only