Learn More
SYPL1 Mouse anti-Human, DyLight 650, Clone: EP10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259340C
Catalog No. NBP259340C
Add to cart
Description
Description
SYPL1 Monoclonal antibody specifically detects SYPL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
SYPL1
EP10
Western Blot, Immunohistochemistry
Purified
SYPL1
Immunoprecipitate of human brain
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 38kDa.
Western Blot, Immunohistochemistry
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL
Mouse
IgG1
100 ul
Primary
6856
Human