Learn More
Invitrogen™ Syrian Hamster IgG Isotype Control, eBioscience™
Syrian Hamster Polyclonal Isotype Control
Supplier: Invitrogen™ 14491481
Description
Description: This hamster IgG fraction was purified by affinity chromatography from normal Golden Syrian hamster serum. Applications Reported: Golden Syrian hamster IgG isotype control has been reported for use in surface and intracellular flow cytometric analysis, immunohistochemical staining, immunoprecipitation and immunoblotting (WB). Applications Tested: This istoype control antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. Use isotype control at the same concentration as experimental antibody. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications
| Syrian Hamster IgG | |
| Polyclonal | |
| Unconjugated | |
| IgG; Immunoglobulin G; ImmunoglobulinG | |
| Affinity chromatography | |
| RUO | |
| Not Applicable | |
| Isotype Control | |
| IgG |
| Control, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Syrian Hamster | |
| 50 μg | |
| Isotype Control (Primary) | |
| 4°C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.