Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Syringe Filters: Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.

Low hold-up volume helps you to recover virtually all of your sample after filtering

 View more versions of this product

Catalog No. 09-720-000


Add to cart

Description

Description

Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.

Nylon Membrane

  • Excellent chemical resistance and low protein binding
  • Ideal for aqueous/organic/HPLC applications
  • Hydrophilic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Non-sterile
13mm
Ideal for aqueous/organic/HPLC applications
0.22μm
100/Pk.
Syringe Filter
Nylon (NYL)
Low
100
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit