Learn More
Azzota Corp Syringe Filter, Nylon, Diameter: 33mm, Pore Size: 0.22um, 50/pk, Individually wrapped, Sterile DFS Item
Manufacturer:  Azzota Corp AF330122S
Syringe Filter, Nylon, Diameter: 33mm, Pore Size: 0.22um, 50/pk, Individually wrapped, Sterile
Catalog No. 50-161-2761