Learn More
Thermo Scientific™ System III Liquid to Liquid Heat Exchanger
Compact, reliable heat exchangers provide a low energy and efficient way to remove heat from water-cooled applications resulting in low cost of ownership
Manufacturer:  Thermo Scientific™ 327005301602
 View more versions of this product
Catalog No. 13-265-238
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pump type: TU9
±1.0°C
Reservoir Volume: 1.25 gal. / 4.7L
+5°C to +45°C
70kW / 238840BTU
Dimensions: 20 3/4 x 17 3/8 x 27 in. (52.7 x 44.1 x 68.6cm)