Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT10 Mouse anti-Human, Mouse, Rat, Clone: S269-73, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527591

Catalog No. PIMA527591


Add to cart

Description

Description

SYT10 is a Ca2+ sensor specifically required for the Ca2+-dependent exocytosis of secretory vesicles containing IGF1 in neurons of the olfactory bulb. Exocytosis of IGF1 is required for sensory perception of smell. SYT10 is not involved in Ca2+-dependent synaptic vesicle exocytosis. It acts through Ca2+ and phospholipid binding to the C2 domain: Ca2+ induces binding of the C2-domains to phospholipid membranes and to assembled SNARE-complexes; both actions contribute to triggering exocytosis.
Specifications

Specifications

SYT10
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O08625, Q6XYQ8, Q9R0N4
SYT10, SYT-10, Synaptotagmin 10, Synaptotagmin X, SytX, P65, SVP65, DKFZp781D2042, SYT
Mouse
IgG1
100 μg
-20°C
Primary
341359, 54526, 60567
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S269-73
Unconjugated
SYT10
Liquid
SYT10
Fusion protein amino acids 248-335 (Cytoplasmic C2A domain) of mouse Synaptotagmin-10
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit