Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551067

Catalog No. PA551067


Add to cart

Description

Description

SYT13 Polyclonal Antibody detects endogenous levels of SYT13 protein.

SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.
Specifications

Specifications

SYT13
Polyclonal
Unconjugated
SYT13
Liquid
SYT13
Synthesized peptide derived from the Internal region of human Synaptotagmin XIII.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q7L8C5, Q9EQT6, Q925B5
KIAA1427
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57586, 80976, 80977
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit