Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549193

Catalog No. PA549193

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in Ca(2+)-dependent exocytosis of secretory vesicles through Ca(2+) and phospholipid binding to the C2 domain or may serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis.
Specifications

Specifications

Synaptotagmin 4
Polyclonal
PBS with 0.09% sodium azide
Q9H2B2
Synaptotagmin-4, Synaptotagmin IV, SytIV, SYT4, KIAA1342
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6860
Western Blot
Unconjugated
SYT4
Liquid
SYT4
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human SYT4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit