Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Syt5, Mouse anti-Mouse, A655, Clone: S276-15, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18487

Catalog No. 89-338-412



Description

Description

Specifications

Specifications

Syt5
S276-15
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
synaptotagmin V
SytV
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
A655
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Syt5
Recombinant protein corresponding to amino acids 253-344 at cytoplasmic C2B domain of mouse Syt5.
100 μg
Store at -20°C.
Primary
53420
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit