Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syt6, Mouse anti-Mouse, ATTO700, Clone: S269-73, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18218

Catalog No. 89-338-143


Add to cart

Description

Description

Specifications

Specifications

Syt6
S269-73
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
synaptotagmin VI
3110037A08Rik|AW048930
Mouse
IgG1
RUO
Antibody
Monoclonal
It does not cross react with synaptotagmin-6 or synaptotagmin-3 or others.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
ATTO700
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Syt6
Recombinant protein corresponding to amino acids 248-335 at cytoplasmic C2A domain of mouse Syt10.
100 μg
Store at -20°C.
Primary
54524
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit