Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550270

Catalog No. PA550270


Add to cart

Description

Description

The antibody detects endogenous levels of total SYT7 protein.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. A similar protein in rodents mediates hormone secretion and lysosome exocytosis. In humans, expression of this gene has been associated with prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

Synaptotagmin 7
Polyclonal
Unconjugated
SYT7
Liquid
SYT7
Fusion protein corresponding to a region derived from internal residues of human synaptotagmin VII
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O43581
PCANAP7
Rabbit
IgG
100 μL
-20°C
Primary
9066
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit