Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYTL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SYTL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179866
|
Novus Biologicals
NBP179866 |
100 μL |
Each of 1 for $436.00
|
|
Description
SYTL1 Polyclonal specifically detects SYTL1 in Human samples. It is validated for Western Blot.Specifications
SYTL1 | |
Polyclonal | |
Rabbit | |
Human | |
NP_116261 | |
84958 | |
Synthetic peptide directed towards the N terminal of human SYTL1. Peptide sequence: EAEAAVKEKEEGPEPRLTIDEAPQERLRETEASDPEEASQAQEDPGQGDQ | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
exophilin-7, FLJ14996, JFC1, NADPH oxidase-related, C2 domain-containing protein, SLP1, synaptotagmin-like 1, synaptotagmin-like protein 1 | |
SYTL1 | |
IgG | |
Affinity Purified | |
61 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title