Learn More
T 0070907, Tocris
Highly potent and selective PPARγ antagonist
Manufacturer:  R&D Systems 2301/10
Quantity 10mg
 View more versions of this product
Catalog No. 23-011-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
277.67
T 0070907
C12H8N3O3Cl
2-Chloro-5-nitro-N-4-pyridinylbenzamide
Specifications
10mg
>99%
277.67
T 0070907
313516-66-4
C12H8N3O3Cl
2-Chloro-5-nitro-N-4-pyridinylbenzamide
Safety and Handling
Recommended Storage : Store at Room Temperature