Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T 0156 hydrochloride, Tocris

Highly potent, selective PDE5 inhibitor

Manufacturer:  R&D Systems 1676/50

Quantity 50mg

 View more versions of this product

Catalog No. 16-765-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>99%
620.06
T 0156 hydrochloride
C31H29N5O7·HCl
1,2-Dihydro-2-[(2-methyl-4-pyridinyl)methyl]-1-oxo-8-(2-pyrimidinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-2,7-naphthyridine-3-carboxylic acid methyl ester hydrochloride

Specifications

50mg
>99%
620.06
T 0156 hydrochloride
324572-93-2
C31H29N5O7·HCl
1,2-Dihydro-2-[(2-methyl-4-pyridinyl)methyl]-1-oxo-8-(2-pyrimidinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-2,7-naphthyridine-3-carboxylic acid methyl ester hydrochloride

Safety and Handling

Recommended Storage : Desiccate at +4°C

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'