Learn More
Cole-Parmer Parker Hannifin FBMT-12124 Flare compression male branch adapter, 3/4" x 1/4", PFA, Each  DFS Item
Manufacturer:  Cole-Parmer 3049097
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
Catalog No. 50-631-876