Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet/TBX21 Mouse anti-Human, Mouse, Rhesus Macaque, PE/Cy5.5, Clone: 4B10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP143298PECY55
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Flow Cytometry.Specifications
T-bet/TBX21 | |
4B10 | |
Flow Cytometry | |
Conjugated, Purified | |
TBX21 | |
The immunogen for this antibody was TBet. | |
Protein A or G purified | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human, Mouse, Rhesus Macaque |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Primary | |
30009.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title