Learn More
Cole-Parmer Masterflex Fitting, PVDF, Tee, Hose Barb Union, 1/4" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3070375
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-665