Learn More
Cole-Parmer Masterflex Fitting, Stainless Steel, Tee, Hose Barb Union, 3/8" ID  DFS Item
Manufacturer:  Cole-Parmer 3062029
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-453