Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
t-PlasminogenActivator/tPA Mouse anti-Human, Clone: AH54-10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267279
Description
t-PlasminogenActivator/tPA Monoclonal antibody specifically detects t-PlasminogenActivator/tPA in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
t-PlasminogenActivator/tPA | |
AH54-10 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.2% Sodium Azide | |
Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator | |
Mouse | |
IgG1 | |
100μL | |
Primary | |
5327 |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
Western Blot 1:1000 ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
PLAT | |
recombinant protein | |
Protein A purified | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title