Learn More
MilliporeSigma™ Supelco™ T-Type Gas Line Filters
Remove dust and small particles that could otherwise foul downstream needle valves
$193.20 - $285.00
Specifications
For Use With (Application) Removing Dust and Small Particles That Could Otherwise Foul Downstream Needle Valves
Includes Replaceable 7 µm Sintered Frit, Nuts, and Ferrules
View More Specs
Products 3
Catalog Number Mfr. No. Type Material Price Quantity & Availability  
Catalog Number Mfr. No. Type Material Price Quantity & Availability  
11-100-9778
View Documents
MilliporeSigma™ Supelco™
25807
1/8 in. Male Stainless Steel
Each for $285.00
Add to Cart
11-100-9819
View Documents
MilliporeSigma™ Supelco™
25808
1/4 in. Male Stainless Steel
Each for $271.00
Add to Cart
11-101-0306
View Documents
MilliporeSigma™ Supelco™
25806
1/4 in. Male Brass
Each for $193.20
Add to Cart
Description
Description
Specifications
Specifications
Removing Dust and Small Particles That Could Otherwise Foul Downstream Needle Valves
Replaceable 7 µm Sintered Frit, Nuts, and Ferrules