Learn More
Morwell MD Biosciences T1/ST2 (IL-33 R) Mouse, Monoclonal Antibody, PE Conjugated, 0.1 mL
SDP
Supplier:  Morwell MD Biosciences 101001PE
Encompass
Mouse T1/ST2 (IL-33 R) PE conjugated monoclonal antibody (Clone DJ8, Host / Isotype Subclass: Rat IgG1, light chain not isotyped) for the identification and purification of murine T helper 2 (Th2) cells and all forms of murine mast cells.
Catalog No. NC0684646