Learn More
T122 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS15558R
Encompass_Preferred
Catalog No. 50-198-3880
Add to cart
Description
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
T122
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IFT122
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
55764
IFT122
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q9HBG6
C86139; CED; CED1; hypothetical protein zgc:77217; IFT122; intraflagellar transport 122; intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas); Intraflagellar transport protein 122 homolog; sopb; SPG; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140; Wdr10; WDR10p; WDR140; zgc:77217
KLH conjugated synthetic peptide derived from human IFT122.
Protein A
RUO
Antibody
Polyclonal
Human