Learn More
Tocris Bioscience T2AA
Proliferating cell nuclear antigen (PCNA) inhibitor
Manufacturer: Tocris Bioscience 4723/10
This item is not returnable.
View return policy
Description
- Inhibits PCNA/PIP-box peptide interaction (IC50 ~1 µM)
- Causes DNA replication stress by stalling DNA replication forks
- Inhibits PCNA interaction with DNA polymerase δ and arrests cell growth in S-phase
- Suppresses cancer cell growth
Specifications
(βS)-β-Amino-4-(4-hydroxyphenoxy)-3,5-diiodobenzenpropanol | |
C15H15I2NO3 | |
10 mg |
1380782-27-3 | |
511.09 | |
0.98 |
Safety and Handling
Recommended Storage : Store at -20°C