Learn More
Tocris Bioscience™ T2AA
Proliferating cell nuclear antigen (PCNA) inhibitor
Manufacturer:  Tocris Bioscience™ 4723/50
 View more versions of this product
Catalog No. 47-235-0
Add to cart
This item is not returnable. View return policy
Description
Description
  • Inhibits PCNA/PIP-box peptide interaction (IC50 ~1 µM)
  • Causes DNA replication stress by stalling DNA replication forks
  • Inhibits PCNA interaction with DNA polymerase δ and arrests cell growth in S-phase
  • Suppresses cancer cell growth
Specifications
Specifications
(βS)-β-Amino-4-(4-hydroxyphenoxy)-3,5-diiodobenzenpropanol
C15H15I2NO3
50 mg
1380782-27-3
511.09
0.98
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C