Learn More
Thermo Scientific™ T3 RNA Polymerase: PROMO
Thermo Scientific™ Fermentas T3 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Manufacturer:  Thermo Scientific™ EP0103
 View more versions of this product
Catalog No. FEREP0103PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from its promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Note:
Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA–T3 TAATACGACTCACTATA G GGAGA–T7 ATTTAGGTGACACTATA G AAGNG–SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).
Specifications
Specifications
200U/μL
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
10,000U
T3 RNA Polymerase
1mL of 5X Transcription Buffer