Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ T4 DNA Ligase: PROMO

T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.

$165.02 - $606.80

Specifications

Inhibitors T4 DNA Ligase is strongly inhibited by NaCl or KCl if the concentration exceeds 200mM.
Molecular Weight 55.3 kDa monomer
Source E.coli cells with a cloned gene 30 of bacteriophage T4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FEREL0011PM View Documents Thermo Scientific™
EL0011
Each for $165.02
Add to cart
 
FEREL0012PM View Documents Thermo Scientific™
EL0012
Each for $606.80
Add to cart
 
Description

Description

T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.

The T4 DNA Ligase requires ATP as a cofactor.

Highlights

  • Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
  • Fast - sticky-end ligation is completed in 10 minutes at room temperature
  • Supplied with PEG solution for efficient blunt-end ligation
Specifications

Specifications

T4 DNA Ligase is strongly inhibited by NaCl or KCl if the concentration exceeds 200mM.
55.3 kDa monomer
E.coli cells with a cloned gene 30 of bacteriophage T4
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit